Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMT
Entrez Gene ID
835044
Gene Name
methionine S-methyltransferase
Gene Aliases
AT5G49810, methionine S-methyltransferase
Chromosome Location
Chr.5: 20239293 - 20246398 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124359.4 NP_199792.1 5 - 6 1035 91
GenBank mRNA AY094459.1 - 5 - 6 985 91
BT002664.1 - 5 - 6 910 91

View Full Product Details