Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AFB5
Entrez Gene ID
835062
Gene Name
auxin F-box protein 5
Gene Aliases
AT5G49980, K9P8.12, K9P8_12, auxin F-box protein 5
Chromosome Location
Chr.5: 20333897 - 20336671 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124377.3 NP_568718.1 2 - 3 1244 76
GenBank mRNA AY056431.1 - 2 - 3 1239 76
AY139774.1 - 2 - 3 1239 76
BT004536.1 - 2 - 3 1104 76

View Full Product Details