Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QS
Entrez Gene ID
835086
Gene Name
quinolinate synthase
Gene Aliases
AT5G50210, K6A12.7, K6A12_7, OLD5, ONSET OF LEAF DEATH 5, SUFE3, SULFUR E 3, quinolinate synthase
Chromosome Location
Chr.5: 20442613 - 20446050 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124400.3 NP_199832.1 2 - 3 1228 62
GenBank mRNA AY035115.1 - 2 - 3 1194 62
AY085553.1 - 2 - 3 1135 62
AY113860.1 - 2 - 3 1038 62

View Full Product Details