Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HVA22E
Entrez Gene ID
835144
Gene Name
HVA22 homologue E
Gene Aliases
ARABIDOPSIS THALIANA HVA22 HOMOLOGUE E, AT5G50720, ATHVA22E, HVA22 homologue E, MFB16.12, MFB16_12
Chromosome Location
Chr.5: 20633193 - 20634618 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124450.5 NP_568744.1 4 - 5 453 76
GenBank mRNA AF290892.1 - 4 - 5 375 76

View Full Product Details