Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EYE
- Entrez Gene ID
-
835217
- Gene Name
- conserved oligomeric Golgi complex component-related / COG complex component-like protein
- Gene Aliases
-
AT5G51430, EMBRYO YELLOW, MFG13.14, MFG13_14
- Chromosome Location
-
Chr.5: 20886745 - 20891007 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124522.4 |
NP_199956.1 |
7 - 8 |
1440 |
82 |
View Full Product Details