Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KEA5
Entrez Gene ID
835245
Gene Name
K+ efflux antiporter 5
Gene Aliases
ARABIDOPSIS THALIANA K+ EFFLUX ANTIPORTER 5, AT5G51710, ATKEA5, K+ efflux antiporter 5, MIO24.16, MIO24_16
Chromosome Location
Chr.5: 21004251 - 21008849 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125943.1 NP_001119415.1 9 - 10 903 127
NM_124550.2 NP_568763.1 9 - 10 1034 127
GenBank mRNA AY070064.1 - 9 - 10 849 127
AY084820.1 - 9 - 10 902 127

View Full Product Details