Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GA20OX2
Entrez Gene ID
835256
Gene Name
gibberellin 20 oxidase 2
Gene Aliases
AT2353, AT5G51810, ATGA20OX2, MIO24.5, MIO24_5, gibberellin 20 oxidase 2
Chromosome Location
Chr.5: 21055179 - 21056881 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344962.1 NP_001332048.1 1 - 2 680 128
NM_124560.4 NP_199994.1 1 - 2 680 128
GenBank mRNA X83380.1 - - 586 128

View Full Product Details