Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHOC1
Entrez Gene ID
835305
Gene Name
shortage in chiasmata 1
Gene Aliases
AT5G52290, K24M7.2, K24M7_2, SHORTAGE IN CHIASMATA 1
Chromosome Location
Chr.5: 21230224 - 21236554 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124608.5 NP_200042.4 5 - 6 2568 85

View Full Product Details