Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EXO70A2
- Entrez Gene ID
-
835310
- Gene Name
- exocyst subunit exo70 family protein A2
- Gene Aliases
-
AT5G52340, ATEXO70A2, K24M7.7, K24M7_7, exocyst subunit exo70 family protein A2
- Chromosome Location
-
Chr.5: 21250646 - 21254137 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124613.4 |
NP_200047.3 |
3 - 4 |
614 |
120 |
View Full Product Details