Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EDA41
- Entrez Gene ID
-
835322
- Gene Name
- FBD, F-box and Leucine Rich Repeat domains containing protein
- Gene Aliases
-
AT5G52460, K24M7.21, K24M7_21, embryo sac development arrest 41
- Chromosome Location
-
Chr.5: 21292375 - 21293758 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124625.2 |
NP_200059.2 |
1 - 2 |
209 |
72 |
View Full Product Details