Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY27
Entrez Gene ID
835360
Gene Name
WRKY DNA-binding protein 27
Gene Aliases
ARABIDOPSIS THALIANA WRKY DNA-BINDING PROTEIN 27, AT5G52830, ATWRKY27, MXC20.5, MXC20_5, WRKY DNA-binding protein 27
Chromosome Location
Chr.5: 21410769 - 21412526 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124661.3 NP_568777.1 1 - 2 667 98
GenBank mRNA AB493790.1 - 1 - 2 440 98
AF418310.1 - 1 - 2 440 98
AK230436.1 - 1 - 2 584 98
BT026451.1 - 1 - 2 440 98

View Full Product Details