Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAP46
Entrez Gene ID
835379
Gene Name
2A phosphatase associated protein of 46 kD
Gene Aliases
2A phosphatase associated protein of 46 kD, AT5G53000, MNB8.6, MNB8_6
Chromosome Location
Chr.5: 21485317 - 21488042 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124679.3 NP_568783.1 4 - 5 582 63
GenBank mRNA AF133708.1 - 4 - 5 525 63
BT004072.1 - 4 - 5 526 63
BT006150.1 - 4 - 5 409 63

View Full Product Details