Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTB2
Entrez Gene ID
835399
Gene Name
polypyrimidine tract-binding protein 2
Gene Aliases
AT5G53180, ATPTB2, MFH8.12, MFH8_12, POLYPYRIMIDINE TRACT-BINDING PROTEIN 2, polypyrimidine tract-binding protein 2
Chromosome Location
Chr.5: 21568131 - 21571904 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085281.2 NP_001078750.1 5 - 6 651 95
NM_001345040.1 NP_001332166.1 4 - 5 803 95
NM_001345041.1 NP_001332168.1 5 - 6 656 95
NM_001345042.1 NP_001332167.1 4 - 5 585 95
NM_001345043.1 NP_001332169.1 1 - 2 506 95
NM_124697.5 NP_200130.1 4 - 5 593 95
GenBank mRNA AK228857.1 - 5 - 6 651 95
BT015760.1 - 4 - 5 519 95
BT020199.1 - 3 - 4 456 95

View Full Product Details