Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDG29
Entrez Gene ID
835424
Gene Name
SET domain group 29
Gene Aliases
AT5G53430, ATX5, MYN8.4, MYN8_4, SET domain group 29, SET29
Chromosome Location
Chr.5: 21677146 - 21684122 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124722.3 NP_200155.2 4 - 5 1707 84
GenBank mRNA AK117221.1 - 4 - 5 1707 84
BT005981.1 - 4 - 5 1230 84

View Full Product Details