Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAMI
Entrez Gene ID
835498
Gene Name
membrane-associated mannitol-induced
Gene Aliases
AT5G54110, ATMAMI, membrane-associated mannitol-induced
Chromosome Location
Chr.5: 21958059 - 21960634 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345079.1 NP_001331145.1 4 - 5 1469 82
NM_124791.3 NP_568804.1 5 - 6 916 82
GenBank mRNA BT025794.1 - 5 - 6 619 82

View Full Product Details