Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MOR1
Entrez Gene ID
818131
Gene Name
ARM repeat superfamily protein
Gene Aliases
AT2G35630, GEM1, MICROTUBULE ORGANIZATION 1, T20F21.17, T20F21_17
Chromosome Location
Chr.2: 14966574 - 14980761 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336567.1 NP_001325149.1 1 - 2 359 63
NM_129117.5 NP_565811.2 1 - 2 420 63
GenBank mRNA AF367246.1 - 1 - 2 166 63

View Full Product Details