Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FIS2
- Entrez Gene ID
-
818136
- Gene Name
- VEFS-Box of polycomb protein
- Gene Aliases
-
AT2G35670, FERTILIZATION INDEPENDENT SEED 2, FERTILIZATION-INDEPENDENT ENDOSPERM 2, FERTILIZATION-INDEPENDENT SEED 2 PROTEIN, FIE2, T20F21.14, T20F21_14
- Chromosome Location
-
Chr.2: 14992565 - 14996756 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129122.3 |
NP_565815.2 |
12 - 13 |
2258 |
104 |
View Full Product Details