Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACX5
Entrez Gene ID
818138
Gene Name
acyl-CoA oxidase 5
Gene Aliases
AT2G35690, T20F21.12, T20F21_12, acyl-CoA oxidase 5
Chromosome Location
Chr.2: 14999726 - 15003073 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129124.4 NP_181112.1 9 - 10 1657 62
GenBank mRNA AY074357.1 - 9 - 10 1629 62
AY096691.1 - 9 - 10 1421 62

View Full Product Details