Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl26
Entrez Gene ID
818150
Gene Name
serine carboxypeptidase-like 26
Gene Aliases
AT2G35780, T20F21.2, serine carboxypeptidase-like 26
Chromosome Location
Chr.2: 15037579 - 15040298 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129136.5 NP_181121.1 1 - 2 547 108
GenBank mRNA AY088044.1 - 1 - 2 367 108
BT029178.1 - 1 - 2 353 108

View Full Product Details