Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
E2F3
Entrez Gene ID
818174
Gene Name
E2F transcription factor 3
Gene Aliases
AT2G36010, ATE2FA, E2F TRANSCRIPTION FACTOR-3 E2F3, E2F transcription factor 3, F11F19.8, F11F19_8
Chromosome Location
Chr.2: 15119516 - 15123184 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129160.4 NP_565831.3 1 - 2 422 64
NM_201882.3 NP_973611.1 1 - 2 448 64
GenBank mRNA AF242582.1 - 1 - 2 685 64
AJ294534.1 - 1 - 2 276 64
BT026376.1 - 1 - 2 276 64

View Full Product Details