Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DML1
Entrez Gene ID
818224
Gene Name
demeter-like 1
Gene Aliases
AT2G36490, AtROS1, F1O11.12, F1O11_12, REPRESSOR OF SILENCING1, ROS1, demeter-like 1
Chromosome Location
Chr.2: 15307907 - 15314807 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129207.5 NP_181190.3 11 - 12 3363 108
GenBank mRNA AY286009.1 - 10 - 11 3070 108

View Full Product Details