Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAB7
Entrez Gene ID
818238
Gene Name
poly(A) binding protein 7
Gene Aliases
AT2G36660, F13K3.6, F13K3_6, poly(A) binding protein 7
Chromosome Location
Chr.2: 15361321 - 15364398 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129221.2 NP_181204.1 7 - 8 1366 101

View Full Product Details