Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHT4;2
Entrez Gene ID
818384
Gene Name
phosphate transporter 4;2
Gene Aliases
AT2G38060, T8P21.3, T8P21_3, phosphate transporter 4;2
Chromosome Location
Chr.2: 15922518 - 15925895 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336686.1 NP_001325125.1 3 - 4 1266 75
NM_129362.4 NP_181341.2 3 - 4 1274 75

View Full Product Details