Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY33
Entrez Gene ID
818429
Gene Name
WRKY DNA-binding protein 33
Gene Aliases
AT2G38470, ATWRKY33, T19C21.4, T19C21_4, WRKY DNA-BINDING PROTEIN 33, WRKY DNA-binding protein 33
Chromosome Location
Chr.2: 16108235 - 16110766 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129404.4 NP_181381.2 4 - 5 1382 63
GenBank mRNA AK226301.1 - 4 - 5 1159 63
BT033030.1 - 4 - 5 1141 63

View Full Product Details