Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CPK20
- Entrez Gene ID
-
818476
- Gene Name
- calcium-dependent protein kinase 20
- Gene Aliases
-
AT2G38910, T7F6.8, T7F6_8, calcium-dependent protein kinase 20
- Chromosome Location
-
Chr.2: 16244906 - 16247855 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129449.2 |
NP_181425.1 |
3 - 4 |
1534 |
100 |
View Full Product Details