Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SVR1
Entrez Gene ID
818500
Gene Name
pseudouridine synthase family protein
Gene Aliases
AT2G39140, AT2G39150, PDE328, PIGMENT DEFECTIVE 328, SUPPRESSOR OF VARIEGATION 1, T16B24.22, T16B24_22
Chromosome Location
Chr.2: 16330129 - 16333405 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336760.1 NP_001323702.1 3 - 4 819 120
NM_129471.3 NP_181447.2 3 - 4 873 120
GenBank mRNA AK117682.1 - 3 - 4 858 120
AY088622.1 - 3 - 4 806 120

View Full Product Details