Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JAL23
Entrez Gene ID
818518
Gene Name
jacalin-related lectin 23
Gene Aliases
AT2G39330, T16B24.3, T16B24_3, jacalin-related lectin 23
Chromosome Location
Chr.2: 16419483 - 16422158 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336766.1 NP_001318382.1 4 - 5 919 102
NM_001336767.1 NP_001323546.1 2 - 3 960 102
NM_129490.4 NP_181465.1 3 - 4 778 102
GenBank mRNA AY087871.1 - 3 - 4 720 102

View Full Product Details