Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXO70H2
Entrez Gene ID
818523
Gene Name
exocyst subunit exo70 family protein H2
Gene Aliases
AT2G39380, ATEXO70H2, F12L6.4, F12L6_4, exocyst subunit exo70 family protein H2
Chromosome Location
Chr.2: 16447006 - 16449083 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129495.2 NP_181470.1 1 - 1 1541 78

Gene Symbol
AT2G39375
Entrez Gene ID
28718337
Gene Name
hypothetical protein
Gene Aliases
AT2G39375
Chromosome Location
Chr.2: 16446514 - 16449180 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336769.1 NP_001323671.1 2 - 2 584 78
NM_001336770.1 NP_001323672.1 2 - 2 524 78
NM_001336771.1 NP_001323673.1 1 - 1 404 78

View Full Product Details