Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGGT-I
Entrez Gene ID
818540
Gene Name
Prenyltransferase family protein
Gene Aliases
AT2G39550, ATGGT-IB, GERANYLGERANYLTRANSFERASE-I BETA SUBUNIT, GGB
Chromosome Location
Chr.2: 16501571 - 16504379 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129513.4 NP_181487.1 6 - 7 748 82
GenBank mRNA BT002465.1 - 6 - 7 696 82
BT008464.1 - 6 - 7 653 82

View Full Product Details