Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNS2
Entrez Gene ID
818563
Gene Name
ribonuclease 2
Gene Aliases
AT2G39780, T5I7.8, T5I7_8, ribonuclease 2
Chromosome Location
Chr.2: 16591117 - 16593775 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125006.1 NP_001118478.1 6 - 7 668 75
NM_129536.5 NP_030524.1 8 - 9 869 75
GenBank mRNA BT025792.1 - 8 - 9 730 75
M98336.1 - 8 - 9 745 75

View Full Product Details