Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ETG1
Entrez Gene ID
818650
Gene Name
E2F target protein 1 (ETG1)
Gene Aliases
AT2G40550, E2F target gene 1, T2P4.10, T2P4_10
Chromosome Location
Chr.2: 16934621 - 16938272 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129617.5 NP_181587.2 8 - 9 1332 72
GenBank mRNA BT022032.1 - 8 - 9 1041 72

View Full Product Details