Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RR16
Entrez Gene ID
818662
Gene Name
response regulator 16
Gene Aliases
ARR16, AT2G40670, RESPONSE REGULATOR 16, T7D17.15, T7D17_15, response regulator 16
Chromosome Location
Chr.2: 16969673 - 16972113 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202793.2 NP_001189722.1 2 - 3 650 98
NM_129629.3 NP_181599.1 2 - 3 831 98
GenBank mRNA AF305721.1 - 2 - 3 246 98
AJ279079.1 - 2 - 3 246 98
BT043482.1 - 2 - 3 246 98

View Full Product Details