Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RR3
Entrez Gene ID
818730
Gene Name
response regulator 3
Gene Aliases
ARR8, AT2G41310, ATRR3, RESPONSE REGULATOR 8, response regulator 3
Chromosome Location
Chr.2: 17221840 - 17224880 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129695.4 NP_181663.1 1 - 2 537 76
GenBank mRNA AB010917.1 - 1 - 2 307 76
AK228346.1 - 1 - 2 477 76
AY074643.1 - 1 - 2 99 76

View Full Product Details