Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RR3
Entrez Gene ID
818730
Gene Name
response regulator 3
Gene Aliases
ARR8, AT2G41310, ATRR3, RESPONSE REGULATOR 8, response regulator 3
Chromosome Location
Chr.2: 17221840 - 17224880 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129695.4 NP_181663.1 5 - 6 1468 99
GenBank mRNA AB010917.1 - 6 - 7 1121 99
AK228346.1 - 5 - 6 1408 99

View Full Product Details