Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARP
Entrez Gene ID
818744
Gene Name
apurinic endonuclease-redox protein
Gene Aliases
AT2G41460, T26J13.5, T26J13_5, apurinic endonuclease-redox protein
Chromosome Location
Chr.2: 17285470 - 17289023 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125028.1 NP_001118500.1 5 - 6 629 142
NM_001336913.1 NP_001323447.1 5 - 6 869 142
NM_001336914.1 NP_001323448.1 4 - 5 858 142
NM_129709.5 NP_181677.1 5 - 6 890 142

View Full Product Details