Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NTRC
- Entrez Gene ID
-
818766
- Gene Name
- NADPH-dependent thioredoxin reductase C
- Gene Aliases
-
AT2G41680, NADPH-dependent thioredoxin reductase C, T32G6.20, T32G6_20
- Chromosome Location
-
Chr.2: 17376155 - 17379059 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129731.6 |
NP_565954.1 |
3 - 4 |
585 |
73 |
View Full Product Details