Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDF1
Entrez Gene ID
818884
Gene Name
protodermal factor 1
Gene Aliases
AT2G42840, F7D19.16, F7D19_16, protodermal factor 1
Chromosome Location
Chr.2: 17826071 - 17827648 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129845.4 NP_181812.1 1 - 2 348 84
GenBank mRNA AF141375.1 - 1 - 2 136 84
AY088094.1 - 1 - 2 184 84
BT024781.1 - 1 - 2 126 84

View Full Product Details