Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BIO2
Entrez Gene ID
818937
Gene Name
Radical SAM superfamily protein
Gene Aliases
AT2G43360, BIOB, BIOTIN AUXOTROPH 2, BIOTIN AUXOTROPH B, BIOTIN SYNTHASE, T1O24.10
Chromosome Location
Chr.2: 18010676 - 18013252 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129897.4 NP_181864.1 2 - 3 511 74
GenBank mRNA BT024899.1 - 2 - 3 388 74
L34413.1 - 2 - 3 448 74
U31806.1 - 2 - 3 445 74

View Full Product Details