Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TGG3
- Entrez Gene ID
-
834891
- Gene Name
- thioglucoside glucosidase 3
- Gene Aliases
-
AT5G48375, BETA GLUCOSIDASE 39, BGLU39, thioglucoside glucosidase 3
- Chromosome Location
-
Chr.5: 19601303 - 19603883 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_148101.1 |
NP_680406.1 |
7 - 8 |
693 |
112 |
View Full Product Details