Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LKP2
- Entrez Gene ID
-
816408
- Gene Name
- LOV KELCH protein 2
- Gene Aliases
-
ADAGIO 2, ADO2, AT2G18915, LOV KELCH protein 2
- Chromosome Location
-
Chr.2: 8194512 - 8197483 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127448.2 |
NP_565444.2 |
1 - 2 |
252 |
117 |
View Full Product Details