Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WRKY35
Entrez Gene ID
818048
Gene Name
WRKY DNA-binding protein 35
Gene Aliases
AT2G34830, AtWRKY35, F19I3.6, F19I3_6, MEE24, WRKY DNA-BINDING PROTEIN 35, WRKY DNA-binding protein 35, maternal effect embryo arrest 24
Chromosome Location
Chr.2: 14693696 - 14696803 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336530.1 NP_001324223.1 2 - 3 1127 150
NM_129036.2 NP_181029.1 2 - 3 702 150
GenBank mRNA AF404863.1 - 2 - 3 702 150
BT029239.1 - 2 - 3 702 150

View Full Product Details