Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARFD1B
Entrez Gene ID
839242
Gene Name
ADP-ribosylation factor D1B
Gene Aliases
ADP-ribosylation factor D1B, AT1G02430, ATARFD1B, T6A9.12, T6A9_12
Chromosome Location
Chr.1: 495050 - 496332 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331338.1 NP_001321619.1 1 - 2 426 149
NM_100124.5 NP_171745.3 1 - 2 426 149

View Full Product Details