Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGLU21
Entrez Gene ID
842944
Gene Name
Glycosyl hydrolase superfamily protein
Gene Aliases
AT1G66270, T6J19.2
Chromosome Location
Chr.1: 24699868 - 24703166 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105298.4 NP_176801.1 12 - 13 1543 168
NM_179517.2 NP_849848.1 12 - 13 1412 168
GenBank mRNA AY045698.1 - 12 - 13 1413 168
BT002684.1 - 12 - 13 1372 168

View Full Product Details