Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VAR2
Entrez Gene ID
817646
Gene Name
FtsH extracellular protease family
Gene Aliases
AT2G30950, F7F1.16, F7F1_16, FTSH2, VARIEGATED 2
Chromosome Location
Chr.2: 13174279 - 13177223 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336310.1 NP_001323510.1 4 - 5 2051 132
NM_001336311.1 NP_001323511.1 4 - 5 2034 132
NM_001336312.1 NP_001323509.1 4 - 5 1984 132
NM_179825.3 NP_850156.1 4 - 5 1853 132
GenBank mRNA AF135189.1 - 4 - 5 1776 132

View Full Product Details