Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPK2
Entrez Gene ID
819084
Gene Name
thiamin pyrophosphokinase 2
Gene Aliases
AT2G44750, F16B22.24, thiamin pyrophosphokinase 2
Chromosome Location
Chr.2: 18451426 - 18452875 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180093.2 NP_850424.1 3 - 4 391 148

View Full Product Details