Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY69
Entrez Gene ID
825040
Gene Name
WRKY DNA-binding protein 69
Gene Aliases
AT3G58710, ATWRKY69, RABIDOPSIS THALIANA WRKY DNA-BINDING PROTEIN 69, WRKY DNA-binding protein 69
Chromosome Location
Chr.3: 21714911 - 21716916 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180689.3 NP_851020.1 1 - 2 411 119
GenBank mRNA AY084233.1 - 1 - 2 316 119
BT029470.1 - 1 - 2 131 119

View Full Product Details