Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- OPR2
- Entrez Gene ID
-
844002
- Gene Name
- 12-oxophytodienoate reductase 2
- Gene Aliases
-
12-OXOPHYTODIENOATE REDUCTASE 2, 12-oxophytodienoate reductase 2, ARABIDOPSIS 12-OXOPHYTODIENOATE REDUCTASE 2, AT1G76690, ATOPR2, F28O16.6, F28O16_6
- Chromosome Location
-
Chr.1: 28778622 - 28780483 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106319.5 |
NP_177795.1 |
2 - 3 |
574 |
132 |
GenBank mRNA |
BT025251.1 |
- |
2 - 3 |
220 |
132 |
View Full Product Details