Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDG20
Entrez Gene ID
821169
Gene Name
SET domain protein 20
Gene Aliases
AT3G03750, F20H23.22, F20H23_22, SET domain protein 20, SUVR3
Chromosome Location
Chr.3: 939900 - 942094 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111246.3 NP_187025.2 1 - 2 663 119
GenBank mRNA AK227335.1 - 1 - 2 663 119
BT008766.1 - 1 - 2 639 119

View Full Product Details