Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBP47C
Entrez Gene ID
841157
Gene Name
RNA-binding protein 47C
Gene Aliases
AT1G47490, ATRBP47C, F16N3.24, F16N3_24, RNA-binding protein 47C
Chromosome Location
Chr.1: 17424473 - 17427475 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103642.3 NP_175180.1 3 - 4 1239 141
GenBank mRNA AF344325.1 - 3 - 4 911 141
AY035180.1 - 3 - 4 1043 141
AY062960.1 - 3 - 4 911 141

View Full Product Details