Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BGLU35
- Entrez Gene ID
-
841572
- Gene Name
- beta glucosidase 35
- Gene Aliases
-
AT1G51470, F5D21.17, F5D21_17, TGG5, THIOGLUCOSIDE GLUCOHYDROLASE 5, beta glucosidase 35
- Chromosome Location
-
Chr.1: 19087384 - 19090523 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104025.4 |
NP_175558.3 |
9 - 10 |
1176 |
91 |
View Full Product Details